Assay Details
| Gene Symbol: Pacrgl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025755.3 | NP_080031.1 | 6 - 7 | 618 | 82 | |
| GenBank mRNA | AK016966.1 | - | 6 - 7 | 667 | 82 | |
| AK018101.1 | - | 6 - 7 | 627 | 82 | ||
| AK032498.1 | - | 5 - 6 | 2366 | 82 | ||
| BC018481.1 | - | 6 - 7 | 618 | 3482169 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pacrgl
Genomic Map