Assay Details
| Gene Symbol: L3hypdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026038.2 | NP_080314.1 | 4 - 5 | 1098 | 65 | |
| GenBank mRNA | AK012954.1 | - | 4 - 5 | 1220 | 65 | |
| AK018449.1 | - | 4 - 5 | 1099 | 65 | ||
| BC004753.1 | - | 4 - 5 | 1158 | 3584353 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530096 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530097 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530091 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530095 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530098 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530092 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530093 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530094 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408989 | BC004753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408989 | AK012954.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408989 | AK018449.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408989 | NM_026038.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408989 | BC004753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408989 | AK012954.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408989 | AK018449.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408989 | NM_026038.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777714 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683821 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129959 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399709 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156416 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345780 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180108 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177609 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107356 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408989 | BC004753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408989 | AK012954.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408989 | AK018449.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408989 | NM_026038.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408989 | BC004753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408989 | AK012954.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408989 | AK018449.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408989 | NM_026038.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530085 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107356 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07661021 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07661011 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177609 | - | - |
| Whole Mouse Genome G4122A | A_51_P408989 | BC004753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408989 | AK012954.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408989 | AK018449.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408989 | NM_026038.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408989 | BC004753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408989 | AK012954.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408989 | AK018449.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408989 | NM_026038.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102018 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107356 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177609 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09856959 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09856945 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09856963 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10048117 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09856960 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530093 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530092 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530094 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530097 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530098 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530091 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530095 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09816971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10061114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09856963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10333774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10096363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10302549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09949158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10061101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10061107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530062 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10400902 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400898 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400899 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400901 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400897 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400903 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424692_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10400896 | - | - |
| Mouse Genome 430 2.0 Array | 1424692_at | - | - |
| Mouse Genome 430 2.0 Array | 1424692_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67217_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67217_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200013022.mm.1 | NM_026038.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013022.mm.1 | NM_026038.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013022.mm.1 | TC1200001899.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200013023.mm.1 | NM_026038.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013023.mm.1 | NM_026038.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013023.mm.1 | TC1200001899.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424692_at | - | - |
| Mouse Expression Set 430 | 1424692_PM_at | - | - |
| Murine Genome U74v2 Set | 166138_r_at | - | - |
| Murine Genome U74v2 Set | 166138_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424692_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424692_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424692_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424692_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 389809 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2760397 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2760397 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002760397 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230505 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230505 | - | - |
| mouse-6 v1.1 (Illumina) | scl42361.4.1_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2760397 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl42361.4.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760397 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1230505 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230505 | - | - |

L3hypdh
Genomic Map