Assay Details
| Gene Symbol: Cmtm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026066.2 | NP_080342.1 | 1 - 2 | 314 | 63 | |
| GenBank mRNA | AK005375.1 | - | 1 - 2 | 269 | 63 | |
| AK013666.1 | - | 1 - 2 | 314 | 63 | ||
| AY241867.1 | - | 1 - 2 | 304 | 63 | ||
| BC049665.1 | - | 1 - 2 | 301 | 6533928 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cmtm5
Genomic Map