Assay Details
| Gene Symbol: Pabpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163836.1 | NP_001157308.1 | 1 | 3059 | 99 | |
| GenBank mRNA | AK014801.1 | - | 1 | 927 | 99 | |
| AK016578.1 | - | 1 | 3052 | 99 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P330353 | AK016578.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P330353 | AK016578.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012286 | BC113177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012286 | AK016578.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012286 | AK029765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012286 | NM_001163836.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P330353 | AK016578.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P330353 | AK016578.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012286 | BC113177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012286 | AK016578.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012286 | AK029765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012286 | NM_001163836.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P330353 | AK016578.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P330353 | AK016578.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13274563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13139029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13262401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13508696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13115453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13508692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13139033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31034442 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1428807_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10447704 | - | - |
| Mouse Genome 430 2.0 Array | 1428807_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428807_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67543_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67543_PM_at | - | - |
| Mouse Expression Set 430 | 1428807_PM_at | - | - |
| Mouse Expression Set 430 | 1428807_at | - | - |
| Murine Genome U74v2 Set | 165436_r_at | - | - |
| Murine Genome U74v2 Set | 165436_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428807_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428807_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 558674 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241281 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241281 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2975779 | - | - |

Pabpc6
Genomic Map