Assay Details
| Gene Symbol: Patl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026251.2 | NP_080527.2 | 3 - 4 | 77 | ||
| GenBank mRNA | AK015195.1 | - | 3 - 4 | 77 | ||
| BC145646.1 | - | 1 - 2 | 181 | 40130949 | 77 | |
| BY714747.1 | - | 3 - 4 | 77 | |||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK015195.1,BC145646.1,NM_026251.2 transcripts.
Set Membership: |
|

Patl2
Genomic Map