Assay Details
| Gene Symbol: Patl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026251.2 | NP_080527.2 | 5 - 6 | 696 | 67 | |
| GenBank mRNA | AK015195.1 | - | 5 - 6 | 697 | 67 | |
| BC145646.1 | - | 3 - 4 | 474 | 40130949 | 67 | |
| DV658440.1 | - | 3 - 4 | 307 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Patl2
Genomic Map