Assay Details
| Gene Symbol: Patl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026251.2 | NP_080527.2 | 16 - 17 | 1948 | 62 | |
| GenBank mRNA | AK010811.1 | - | 9 - 10 | 1016 | 62 | |
| AK015195.1 | - | 16 - 17 | 1952 | 62 | ||
| BC145646.1 | - | 14 - 15 | 1729 | 40130949 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Patl2
Genomic Map