Assay Details
| Gene Symbol: Lrp2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026278.3 | NP_080554.1 | 3 - 4 | 427 | 109 | |
| GenBank mRNA | AK007202.1 | - | 3 - 4 | 419 | 109 | |
| AK015594.1 | - | 3 - 4 | 109 | |||
| AK016626.1 | - | 432 | 109 | |||
| BC051150.1 | - | 3 - 4 | 427 | 6772814 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrp2bp
Genomic Map