Assay Details
| Gene Symbol: Ccny | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026484.3 | NP_080760.2 | 7 - 8 | 1056 | 117 | |
| GenBank mRNA | AK017493.1 | - | 7 - 8 | 888 | 117 | |
| AK028696.1 | - | 7 - 8 | 1057 | 117 | ||
| AK076556.1 | - | 7 - 8 | 586 | 117 | ||
| BC023321.1 | - | 8 - 9 | 974 | 3995040 | 117 | |
| BC035524.1 | - | 6 - 7 | 811 | 4974858 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccny
Genomic Map