Assay Details
| Gene Symbol: Ubxn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026534.2 | NP_080810.2 | 1 - 2 | 110 | 153 | |
| GenBank mRNA | AB120715.1 | - | 1 - 2 | 103 | 153 | |
| AK012822.1 | - | 1 - 2 | 109 | 153 | ||
| AK013985.1 | - | 1 - 2 | 105 | 152 | ||
| AK028268.1 | - | 1 - 2 | 113 | 153 | ||
| AK032332.1 | - | 1 - 2 | 110 | 153 | ||
| AK049018.1 | - | 1 - 2 | 118 | 153 | ||
| BC076632.1 | - | 1 - 2 | 110 | 30637967 | 153 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB120715.1,AK012822.1,AK028268.1,AK032332.1,AK049018.1,BC076632.1,NM_026534.2 transcripts.
Set Membership: |
|

Ubxn2b
Genomic Map