Assay Details
| Gene Symbol: Haghl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271433.1 | NP_001258362.1 | 6 - 7 | 748 | 64 | |
| NM_001271434.1 | NP_001258363.1 | 6 - 7 | 748 | 64 | ||
| NM_001271435.1 | NP_001258364.1 | 6 - 7 | 748 | 64 | ||
| NM_001271436.1 | NP_001258365.1 | 5 - 6 | 562 | 64 | ||
| NM_001271437.1 | NP_001258366.1 | 5 - 6 | 557 | 64 | ||
| NM_026897.3 | NP_081173.1 | 5 - 6 | 733 | 64 | ||
| GenBank mRNA | AK005274.1 | - | 5 - 6 | 729 | 64 | |
| AK012748.1 | - | 6 - 7 | 745 | 64 | ||
| AK032999.1 | - | 3 - 4 | 706 | 64 | ||
| AK034798.1 | - | 6 - 7 | 723 | 64 | ||
| AK047517.1 | - | 6 - 7 | 703 | 64 | ||
| AK079632.1 | - | 3 - 4 | 897 | 64 | ||
| AK154248.1 | - | 5 - 6 | 538 | 64 | ||
| BC005586.1 | - | 6 - 7 | 694 | 64 | ||
| BC030466.1 | - | 6 - 7 | 733 | 5253213 | 64 | |
| BC083322.1 | - | 6 - 7 | 687 | 6820885 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Haghl
Genomic Map