Assay Details
| Gene Symbol: Nupr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026916.3 | NP_081192.2 | 1 - 2 | 384 | 93 | |
| GenBank mRNA | AK007417.1 | - | 1 - 2 | 359 | 93 | |
| AK016320.1 | - | 1 - 2 | 379 | 93 | ||
| BC100444.1 | - | 1 - 2 | 384 | 93 | ||
| EL963130.1 | - | 1 - 2 | 277 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24091171 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24091173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24091168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24091170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24091172 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113486 | AK148362.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113486 | AK135384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113486 | BC100444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113486 | CJ051706.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113486 | NM_026916.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P324754 | BC100444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P324754 | EL963130.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P324754 | NM_026916.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P324754 | AK007417.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P324754 | AK016320.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P324754 | BC100444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P324754 | EL963130.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P324754 | NM_026916.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P324754 | AK007417.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P324754 | AK016320.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654024 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169370 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141566 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113486 | AK148362.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113486 | AK135384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113486 | BC100444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113486 | CJ051706.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113486 | NM_026916.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P324754 | BC100444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P324754 | EL963130.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P324754 | NM_026916.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P324754 | AK007417.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P324754 | AK016320.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P324754 | BC100444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P324754 | EL963130.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P324754 | NM_026916.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P324754 | AK007417.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P324754 | AK016320.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997691 | AK135384.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24091176 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141566 | - | - |
| Whole Mouse Genome G4122A | A_52_P324754 | BC100444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P324754 | EL963130.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P324754 | NM_026916.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P324754 | AK007417.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P324754 | AK016320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P324754 | BC100444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P324754 | EL963130.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P324754 | NM_026916.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P324754 | AK007417.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P324754 | AK016320.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106658 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123708 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141566 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24091170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24091172 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24091171 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24091173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24091168 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526105 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526104 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444382_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429551_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526101 | - | - |
| Mouse Genome 430 2.0 Array | 1429551_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444382_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444382_at | - | - |
| Mouse Genome 430 2.0 Array | 1429551_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69034_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69034_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500013716.mm.1 | NM_026916.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013716.mm.1 | NM_026916.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013716.mm.1 | TC0500001516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013719.mm.1 | NM_026916.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013719.mm.1 | NM_026916.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013719.mm.1 | TC0500001516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013718.mm.1 | NM_026916.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013718.mm.1 | NM_026916.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013718.mm.1 | TC0500001516.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013715.mm.1 | NM_026916.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013715.mm.1 | NM_026916.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013715.mm.1 | TC0500001516.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429551_PM_at | - | - |
| Mouse Expression Set 430 | 1429551_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429551_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444382_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429551_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444382_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 865642 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4780300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4780300 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004780300 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214985 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214985 | - | - |
| mouse-6 v1.1 (Illumina) | scl27174.3.1_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4780300 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27174.3.1_17-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780300 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1214985 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214985 | - | - |

Nupr1l
Genomic Map