Assay Details
| Gene Symbol: Dcps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027030.2 | NP_081306.1 | 4 - 5 | 647 | 65 | |
| GenBank mRNA | AK005584.1 | - | 4 - 5 | 648 | 65 | |
| AK078253.1 | - | 4 - 5 | 676 | 65 | ||
| AK169641.1 | - | 4 - 5 | 655 | 65 | ||
| AK171018.1 | - | 4 - 5 | 639 | 65 | ||
| AY040775.1 | - | 4 - 5 | 660 | 65 | ||
| BC016273.1 | - | 4 - 5 | 667 | 4511427 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dcps
Genomic Map