Assay Details
| Gene Symbol: Apitd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027263.2 | NP_081539.1 | 2 - 3 | 250 | 65 | |
| GenBank mRNA | AK011725.1 | - | 2 - 3 | 199 | 65 | |
| AK012995.1 | - | 2 - 3 | 232 | 65 | ||
| AK013038.1 | - | 2 - 3 | 232 | 65 | ||
| BC138926.1 | - | 2 - 3 | 191 | 8861948 | 65 | |
| BC138927.1 | - | 2 - 3 | 191 | 8861949 | 65 | |
| DV656255.1 | - | 2 - 3 | 250 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012995.1, AK013038.1 transcripts.
Set Membership: |
|

Apitd1
Genomic Map