Assay Details
| Gene Symbol: Meaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290701.1 | NP_001277630.1 | 1 - 2 | 133 | 89 | |
| NM_027310.4 | NP_081586.1 | 1 - 2 | 133 | 89 | ||
| GenBank mRNA | AK009177.1 | - | 1 - 2 | 121 | 89 | |
| BC061024.1 | - | 1 - 2 | 103 | 89 | ||
| BC108404.1 | - | 1 - 2 | 104 | 30135798 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Meaf6
Genomic Map