Assay Details
| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027342.1 | NP_081618.1 | 2 - 3 | 165 | 54 | |
| GenBank mRNA | AK009946.1 | - | 2 - 3 | 165 | 54 | |
| BC010826.1 | - | 2 - 3 | 174 | 4219531 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam162a
Genomic Map