Assay Details
| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027342.1 | NP_081618.1 | 3 - 4 | 275 | 66 | |
| GenBank mRNA | AK009946.1 | - | 3 - 4 | 275 | 66 | |
| BC010826.1 | - | 3 - 4 | 284 | 4219531 | 66 | |
| M97159.1 | - | 246 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Fam162a
Genomic Map