Assay Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027547.2 | NP_081823.2 | 11 - 12 | 1495 | 69 | |
| GenBank mRNA | AK014501.1 | - | 11 - 12 | 1419 | 69 | |
| AK018310.1 | - | 7 - 8 | 881 | 69 | ||
| BC138901.1 | - | 11 - 12 | 1469 | 8861923 | 69 | |
| BC138902.1 | - | 11 - 12 | 1469 | 8861924 | 69 | |
| BG070775.2 | - | 4 - 5 | 69 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Prdm5
Genomic Map