Assay Details
| Gene Symbol: Fam71d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027597.4 | NP_081873.2 | 5 - 6 | 1255 | 145 | |
| NM_029069.1 | NP_083345.1 | 5 - 6 | 1260 | 145 | ||
| GenBank mRNA | AB473726.1 | - | 3 - 4 | 935 | 145 | |
| AK014851.1 | - | 5 - 6 | 1261 | 145 | ||
| AK015802.1 | - | 5 - 6 | 1107 | 145 | ||
| BC051149.1 | - | 5 - 6 | 1115 | 6707726 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam71d
Genomic Map