Assay Details
| Gene Symbol: Fam71d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029069.1 | NP_083345.1 | 7 - 8 | 1530 | 78 | |
| GenBank mRNA | AK014851.1 | - | 7 - 8 | 1531 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam71d
Genomic Map