Assay Details
| Gene Symbol: Enkur | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027728.2 | NP_082004.1 | 4 - 5 | 811 | 78 | |
| GenBank mRNA | AK017056.1 | - | 4 - 5 | 812 | 78 | |
| AY454124.1 | - | 4 - 5 | 809 | 78 | ||
| BC132425.1 | - | 4 - 5 | 672 | 40130702 | 78 | |
| BC132451.1 | - | 4 - 5 | 672 | 40130728 | 78 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21054132 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21054133 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131164 | NM_027728.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131164 | AK017056.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131164 | AY454124.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131164 | NM_027728.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131164 | AK017056.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131164 | AY454124.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381519 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781296 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27929 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235890 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119098 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566758 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 70361 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365936 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125438 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35473 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111810 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152356 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131164 | NM_027728.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131164 | AK017056.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131164 | AY454124.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131164 | NM_027728.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131164 | AK017056.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131164 | AY454124.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131164 | NM_027728.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131164 | AK017056.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131164 | AY454124.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131164 | NM_027728.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131164 | AK017056.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131164 | AY454124.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01955023 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01955021 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01955028 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01955032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21054133 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21054132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21053961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21054052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01535940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01955042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01417057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01535939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01500170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01955021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01388181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01500186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01535947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01535941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01500166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01955023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01388185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01184070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01388186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01616699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01500193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01493013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21053988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21054087 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10480420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480417 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480415 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480416 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480418 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428960_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10480414 | - | - |
| Mouse Genome 430 2.0 Array | 1428960_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428960_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71233_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71233_at | - | - |
| Mouse Expression Set 430 | 1428960_PM_at | - | - |
| Mouse Expression Set 430 | 1428960_at | - | - |
| Murine Genome U74v2 Set | 165633_at | - | - |
| Murine Genome U74v2 Set | 165633_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428960_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428960_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216601 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216601 | - | - |

Enkur
Genomic Map