Assay Details
| Gene Symbol: Afmid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027827.3 | NP_082103.1 | 4 - 5 | 330 | 75 | |
| GenBank mRNA | AF399717.1 | - | 4 - 5 | 325 | 75 | |
| AK018563.1 | - | 2 - 3 | 266 | 75 | ||
| AY099479.1 | - | 4 - 5 | 325 | 75 | ||
| BC024452.1 | - | 4 - 5 | 301 | 75 | ||
| BC043309.1 | - | 4 - 5 | 296 | 75 | ||
| BC066780.1 | - | 4 - 5 | 309 | 30301832 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afmid
Genomic Map