Assay Details
| Gene Symbol: Afmid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027827.3 | NP_082103.1 | 7 - 8 | 590 | 64 | |
| GenBank mRNA | AF399717.1 | - | 7 - 8 | 585 | 64 | |
| AK018563.1 | - | 5 - 6 | 526 | 64 | ||
| AY099479.1 | - | 7 - 8 | 585 | 64 | ||
| BC024452.1 | - | 7 - 8 | 561 | 64 | ||
| BC043309.1 | - | 7 - 8 | 556 | 64 | ||
| BC052507.1 | - | 3 - 4 | 214 | 64 | ||
| BC066780.1 | - | 7 - 8 | 569 | 30301832 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afmid
Genomic Map