Assay Details
| Gene Symbol: Lrwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027891.4 | NP_082167.2 | 8 - 9 | 1149 | 65 | |
| GenBank mRNA | AK004717.1 | - | 6 - 7 | 2111 | 65 | |
| AK051910.1 | - | 8 - 9 | 1097 | 65 | ||
| AK084948.1 | - | 8 - 9 | 1149 | 65 | ||
| AK153976.1 | - | 8 - 9 | 1120 | 65 | ||
| AK154352.1 | - | 8 - 9 | 1091 | 65 | ||
| AK167020.1 | - | 8 - 9 | 1098 | 65 | ||
| AK172708.1 | - | 8 - 9 | 1117 | 65 | ||
| BC048397.1 | - | 8 - 9 | 1128 | 6396815 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrwd1
Genomic Map