Assay Details
| Gene Symbol: Lrwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027891.4 | NP_082167.2 | 10 - 11 | 1429 | 91 | |
| GenBank mRNA | AK004717.1 | - | 8 - 9 | 2391 | 91 | |
| AK051910.1 | - | 10 - 11 | 1377 | 91 | ||
| AK084948.1 | - | 10 - 11 | 1429 | 91 | ||
| AK153976.1 | - | 10 - 11 | 1400 | 91 | ||
| AK154352.1 | - | 10 - 11 | 1371 | 91 | ||
| AK167020.1 | - | 10 - 11 | 1378 | 91 | ||
| AK172708.1 | - | 10 - 11 | 1397 | 91 | ||
| AK207344.1 | - | 2 - 3 | 136 | 91 | ||
| AK218154.1 | - | 2 - 3 | 289 | 91 | ||
| BC048397.1 | - | 10 - 11 | 1408 | 6396815 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrwd1
Genomic Map