Assay Details
| Gene Symbol: Lrwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027891.4 | NP_082167.2 | 13 - 14 | 1821 | 68 | |
| GenBank mRNA | AK004717.1 | - | 11 - 12 | 2783 | 68 | |
| AK084948.1 | - | 13 - 14 | 1821 | 68 | ||
| AK085327.1 | - | 2 - 3 | 1852 | 68 | ||
| AK153976.1 | - | 13 - 14 | 1792 | 68 | ||
| AK154352.1 | - | 13 - 14 | 1764 | 68 | ||
| AK167020.1 | - | 13 - 14 | 1770 | 68 | ||
| AK167868.1 | - | 1 - 2 | 170 | 68 | ||
| AK172708.1 | - | 13 - 14 | 1790 | 68 | ||
| BC048397.1 | - | 13 - 14 | 1800 | 6396815 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrwd1
Genomic Map