Assay Details
| Gene Symbol: 1300017J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027918.2 | NP_082194.1 | 11 - 12 | 1401 | 107 | |
| GenBank mRNA | AK005035.1 | - | 11 - 12 | 1402 | 107 | |
| BC021390.1 | - | 11 - 12 | 1397 | 5054320 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

1300017J02Rik
Genomic Map