Assay Details
| Gene Symbol: Tars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163617.1 | NP_001157089.1 | 6 - 7 | 847 | 80 | |
| NM_001163618.1 | NP_001157090.1 | 4 - 5 | 604 | 80 | ||
| NM_001163619.1 | NP_001157091.1 | 6 - 7 | 847 | 80 | ||
| NM_027931.3 | NP_082207.2 | 6 - 7 | 847 | 80 | ||
| GenBank mRNA | AK011540.1 | - | 6 - 7 | 733 | 80 | |
| AK080690.1 | - | 6 - 7 | 81 | |||
| AK088675.1 | - | 5 - 6 | 597 | 80 | ||
| AK142685.1 | - | 6 - 7 | 742 | 80 | ||
| AK164589.1 | - | 6 - 7 | 754 | 80 | ||
| AK164825.1 | - | 6 - 7 | 705 | 80 | ||
| AK168075.1 | - | 4 - 5 | 498 | 80 | ||
| AK199920.1 | - | 2 - 3 | 108 | 80 | ||
| BC008995.1 | - | 6 - 7 | 732 | 3157108 | 80 | |
| BC147501.1 | - | 6 - 7 | 731 | 9056383 | 80 | |
| BG087111.2 | - | 4 - 5 | 456 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tars2
Genomic Map