| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300019278.mm.1 |
NM_001163617.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019278.mm.1 |
NM_001163617.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019278.mm.1 |
NM_001163618.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019278.mm.1 |
NM_001163618.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019278.mm.1 |
NM_027931.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019278.mm.1 |
NM_027931.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019278.mm.1 |
TC0300002483.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300019277.mm.1 |
NM_001163617.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019277.mm.1 |
NM_001163617.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019277.mm.1 |
NM_001163618.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019277.mm.1 |
NM_001163618.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019277.mm.1 |
NM_027931.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019277.mm.1 |
NM_027931.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019277.mm.1 |
TC0300002483.mm.1 |
MTA TC |