Assay Details
| Gene Symbol: Serpinb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027971.2 | NP_082247.1 | 1 - 2 | 112 | 82 | |
| GenBank mRNA | AK009018.1 | - | 1 - 2 | 113 | 82 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK009018.1,NM_027971.2 transcripts.
Set Membership: |
|

Serpinb12
Genomic Map