Assay Details
| Gene Symbol: Tmco6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028036.3 | NP_082312.3 | 4 - 5 | 596 | 63 | |
| GenBank mRNA | AK010487.1 | - | 4 - 5 | 523 | 63 | |
| AK046207.1 | - | 4 - 5 | 634 | 63 | ||
| BC005637.1 | - | 4 - 5 | 598 | 3156734 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010487.1 transcripts.
Set Membership: |
|

Tmco6
Genomic Map