| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000023558.mm.1 |
NM_001136237.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023558.mm.1 |
NM_001136237.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023558.mm.1 |
TC1000003143.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023550.mm.1 |
NM_001136237.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023550.mm.1 |
NM_001136237.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023550.mm.1 |
NM_028051.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023550.mm.1 |
NM_028051.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023550.mm.1 |
NM_028092.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023550.mm.1 |
NM_028092.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023550.mm.1 |
TC1000003143.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023552.mm.1 |
NM_028051.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023552.mm.1 |
NM_028051.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023552.mm.1 |
TC1000003143.mm.1 |
MTA TC |