Assay Details
| Gene Symbol: Cyb5r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028057.2 | NP_082333.1 | 2 - 3 | 213 | 58 | |
| GenBank mRNA | AK005159.1 | - | 2 - 3 | 214 | 58 | |
| AK049717.1 | - | 1 - 2 | 435 | 58 | ||
| AK159663.1 | - | 2 - 3 | 183 | 58 | ||
| AK167178.1 | - | 2 - 3 | 207 | 58 | ||
| BC016266.1 | - | 2 - 3 | 422 | 4924006 | 58 | |
| BC024618.1 | - | 2 - 3 | 191 | 4235599 | 58 | |
| JX420288.1 | - | 293 | 58 | |||
| JX420289.1 | - | 2 - 3 | 648 | 58 | ||
| JX420290.1 | - | 2 - 3 | 390 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r1
Genomic Map