Assay Details
| Gene Symbol: Cyb5r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028057.2 | NP_082333.1 | 3 - 4 | 291 | 62 | |
| GenBank mRNA | AK005159.1 | - | 3 - 4 | 292 | 62 | |
| AK049717.1 | - | 2 - 3 | 513 | 62 | ||
| AK159663.1 | - | 3 - 4 | 261 | 62 | ||
| AK167178.1 | - | 3 - 4 | 285 | 62 | ||
| BC016266.1 | - | 3 - 4 | 500 | 4924006 | 62 | |
| BC024618.1 | - | 3 - 4 | 269 | 4235599 | 62 | |
| JX420288.1 | - | 371 | 62 | |||
| JX420290.1 | - | 3 - 4 | 468 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r1
Genomic Map