Assay Details
| Gene Symbol: Ccdc71l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162903.1 | NP_001156375.1 | 1 | 1113 | 68 | |
| GenBank mRNA | AK008367.1 | - | 1 | 257 | 68 | |
| AK171109.1 | - | 1 | 763 | 68 | ||
| BC028641.1 | - | 2 | 406 | 68 | ||
| BC049684.1 | - | 1 | 153 | 68 | ||
| BC061478.1 | - | 1 | 484 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P230624 | AK171109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P230624 | BC028641.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P230624 | BC037374.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P230624 | BC049684.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P230624 | NM_001162903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P230624 | AK171109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P230624 | BC028641.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P230624 | BC037374.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P230624 | BC049684.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P230624 | NM_001162903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598835 | BC061478.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598835 | NM_001162903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598835 | BC061478.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P598835 | NM_001162903.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372089 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639455 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 321937 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129116 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167545 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 98042 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121557 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50690 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128011 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168850 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132240 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144607 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598835 | BC061478.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598835 | NM_001162903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598835 | BC061478.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P598835 | NM_001162903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P230624 | AK171109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P230624 | BC028641.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P230624 | BC037374.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P230624 | BC049684.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P230624 | NM_001162903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P230624 | AK171109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P230624 | BC028641.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P230624 | BC037374.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P230624 | BC049684.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P230624 | NM_001162903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230624 | AK171109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230624 | BC028641.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230624 | BC037374.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230624 | BC049684.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230624 | NM_001162903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230624 | AK171109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230624 | BC028641.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230624 | BC037374.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230624 | BC049684.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230624 | NM_001162903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598835 | BC061478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598835 | NM_001162903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598835 | BC061478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598835 | NM_001162903.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10194781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10194783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10246891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10194801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09793260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10186823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09793246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10029373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28336363 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10395255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395254 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429100_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436839_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395252 | - | - |
| Mouse Genome 430 2.0 Array | 1436839_at | - | - |
| Mouse Genome 430 2.0 Array | 1429100_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436839_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429100_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72123_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72123_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200002160.mm.1 | NM_001162903.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200002160.mm.1 | NM_001162903.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200002160.mm.1 | TC1200000278.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436839_at | - | - |
| Mouse Expression Set 430 | 1436839_PM_at | - | - |
| Mouse Expression Set 430 | 1429100_at | - | - |
| Mouse Expression Set 430 | 1429100_PM_at | - | - |
| Murine Genome U74v2 Set | 102987_PM_at | - | - |
| Murine Genome U74v2 Set | 102987_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436839_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436839_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436839_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436839_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429100_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429100_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 545120 | - | - |
| ABI Mouse Genome Survey Microarray | 584961 | - | - |

Ccdc71l
Genomic Map