Assay Details
| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028250.3 | NP_082526.2 | 3 - 4 | 899 | 84 | |
| GenBank mRNA | AK011781.1 | - | 3 - 4 | 892 | 84 | |
| AK167959.1 | - | 4 - 5 | 839 | 83 | ||
| AK209754.1 | - | 3 - 4 | 214 | 84 | ||
| BC115580.1 | - | 3 - 4 | 491 | 40102241 | 84 | |
| BC115581.1 | - | 3 - 4 | 491 | 40102242 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acbd6
Genomic Map