Assay Details
| Gene Symbol: Ppp2r2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028392.3 | NP_082668.1 | 7 - 8 | 1199 | 70 | |
| NR_073583.1 | 7 - 8 | 1651 | 70 | |||
| GenBank mRNA | AF512670.1 | - | 7 - 8 | 905 | 70 | |
| AF536771.1 | - | 7 - 8 | 968 | 70 | ||
| AK013600.1 | - | 7 - 8 | 1201 | 70 | ||
| AK018119.1 | - | 7 - 8 | 1162 | 70 | ||
| AK039592.1 | - | 7 - 8 | 1042 | 70 | ||
| AK079059.1 | - | 1649 | 70 | |||
| AK148956.1 | - | 7 - 8 | 1170 | 70 | ||
| BC026686.1 | - | 7 - 8 | 1137 | 5065516 | 70 | |
| BC066007.1 | - | 7 - 8 | 1440 | 70 | ||
| BC085147.1 | - | 7 - 8 | 1352 | 70 | ||
| BC088979.1 | - | 7 - 8 | 1134 | 5690600 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r2b
Genomic Map