Assay Details
| Gene Symbol: Ccser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027045.1 | NP_081321.1 | 1 - 2 | 224 | 125 | |
| GenBank mRNA | AK013689.1 | - | 1 - 2 | 178 | 125 | |
| AK147383.1 | - | 1 - 2 | 219 | 125 | ||
| AK172153.1 | - | 1 - 2 | 198 | 125 | ||
| BB871809.1 | - | 1 - 2 | 238 | 125 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK147383.1,AK172153.1,NM_027045.1 transcripts.
Set Membership: |
|

Ccser2
Genomic Map