Assay Details
| Gene Symbol: Trmt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028604.2 | NP_082880.2 | 6 - 7 | 607 | 134 | |
| GenBank mRNA | AF532979.1 | - | 6 - 7 | 607 | 134 | |
| AK010722.1 | - | 6 - 7 | 549 | 134 | ||
| AK014181.1 | - | 5 - 6 | 455 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trmt11
Genomic Map