Assay Details
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166506.1 | NP_001159978.1 | 3 - 4 | 428 | 80 | |
| NM_001166507.1 | NP_001159979.1 | 3 - 4 | 428 | 80 | ||
| NM_028777.3 | NP_083053.2 | 3 - 4 | 428 | 80 | ||
| GenBank mRNA | AK004818.1 | - | 3 - 4 | 353 | 80 | |
| AK047396.1 | - | 3 - 4 | 433 | 80 | ||
| BB646963.1 | - | 3 - 4 | 428 | 80 | ||
| BC005766.1 | - | 3 - 4 | 386 | 2649054 | 80 | |
| BC086620.1 | - | 3 - 4 | 362 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l1
Genomic Map