Assay Details
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166506.1 | NP_001159978.1 | 6 - 7 | 918 | 132 | |
| NM_001166507.1 | NP_001159979.1 | 6 - 7 | 918 | 132 | ||
| NM_028777.3 | NP_083053.2 | 6 - 7 | 918 | 132 | ||
| GenBank mRNA | AK004818.1 | - | 6 - 7 | 843 | 132 | |
| AK047396.1 | - | 6 - 7 | 924 | 133 | ||
| AK172947.1 | - | 3 - 4 | 453 | 132 | ||
| AK213489.1 | - | 1 - 2 | 233 | 133 | ||
| BC005766.1 | - | 6 - 7 | 876 | 2649054 | 132 | |
| BC086620.1 | - | 6 - 7 | 852 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l1
Genomic Map