Assay Details
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166506.1 | NP_001159978.1 | 7 - 8 | 1026 | 91 | |
| NM_001166507.1 | NP_001159979.1 | 7 - 8 | 1026 | 91 | ||
| NM_028777.3 | NP_083053.2 | 7 - 8 | 1026 | 91 | ||
| GenBank mRNA | AK004818.1 | - | 7 - 8 | 951 | 91 | |
| AK047396.1 | - | 7 - 8 | 92 | |||
| AK172947.1 | - | 4 - 5 | 561 | 91 | ||
| AK213489.1 | - | 2 - 3 | 341 | 91 | ||
| BC005766.1 | - | 7 - 8 | 984 | 2649054 | 91 | |
| BC086620.1 | - | 7 - 8 | 960 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l1
Genomic Map