Assay Details
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166506.1 | NP_001159978.1 | 11 - 12 | 1549 | 93 | |
| NM_001166507.1 | NP_001159979.1 | 11 - 12 | 1549 | 93 | ||
| NM_028777.3 | NP_083053.2 | 11 - 12 | 1549 | 93 | ||
| GenBank mRNA | AK004818.1 | - | 11 - 12 | 1474 | 93 | |
| AK047396.1 | - | 11 - 12 | 1557 | 93 | ||
| AK172947.1 | - | 8 - 9 | 1084 | 93 | ||
| AK193303.1 | - | 2 - 3 | 228 | 93 | ||
| BC005766.1 | - | 11 - 12 | 1507 | 2649054 | 93 | |
| BC086620.1 | - | 11 - 12 | 1483 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l1
Genomic Map