Assay Details
| Gene Symbol: Rab10os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015551.1 | 1 - 2 | 141 | 100 | ||
| GenBank mRNA | AK005891.1 | - | 1 - 2 | 421 | 100 | |
| AK007702.1 | - | 1 - 2 | 142 | 100 | ||
| AK171768.1 | - | 1 - 2 | 421 | 100 | ||
| BC038315.1 | - | 1 - 2 | 92 | 100 | ||
| BC099424.1 | - | 1 - 2 | 404 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0600000319.rn
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322123 | BC080728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322123 | AK007702.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322123 | NR_015551.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322123 | BC080728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322123 | AK007702.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322123 | NR_015551.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095354 | AK005891.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095354 | AK171768.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136813 | BC099424.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136813 | BC062897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136813 | AK005891.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136813 | AK171768.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136813 | BC038315.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07520697 | - | - |
| Whole Mouse Genome G4122A | A_52_P361338 | AK005891.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P361338 | BC060685.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28194141 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10394153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394156 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394148 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394145 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394157 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394149 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394152 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431818_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453294_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10394144 | - | - |
| Mouse Genome 430 2.0 Array | 1453294_at | - | - |
| Mouse Genome 430 2.0 Array | 1431818_at | - | - |
| Mouse Genome 430 2.0 Array | 1453294_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431818_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200000026.mm.1 | NR_015551.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200000026.mm.1 | NR_015551.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200000026.mm.1 | TC1200000005.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200000021.mm.1 | NR_015551.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200000021.mm.1 | NR_015551.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200000021.mm.1 | TC1200000005.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200000020.mm.1 | NR_015551.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200000020.mm.1 | NR_015551.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200000020.mm.1 | TC1200000005.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453294_PM_at | - | - |
| Mouse Expression Set 430 | 1431818_at | - | - |
| Mouse Expression Set 430 | 1431818_PM_at | - | - |
| Mouse Expression Set 430 | 1453294_at | - | - |
| Murine Genome U74v2 Set | 95924_at | - | - |
| Murine Genome U74v2 Set | 95924_PM_at | - | - |
| Murine Genome U74v2 Set | 171085_at | - | - |
| Murine Genome U74v2 Set | 171085_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431818_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453294_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431818_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453294_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 686931 | - | - |
| ABI Mouse Genome Survey Microarray | 809934 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 580605 | - | - |

Rab10os
Genomic Map