Assay Details
| Gene Symbol: Exoc3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289487.1 | NP_001276416.1 | 4 - 5 | 934 | 60 | |
| NM_001289488.1 | NP_001276417.1 | 4 - 5 | 685 | 60 | ||
| NM_001289489.1 | NP_001276418.1 | 3 - 4 | 580 | 60 | ||
| NM_028807.4 | NP_083083.3 | 3 - 4 | 829 | 60 | ||
| GenBank mRNA | AK004674.1 | - | 4 - 5 | 686 | 60 | |
| AK005443.1 | - | 2 - 3 | 576 | 60 | ||
| AK154875.1 | - | 4 - 5 | 1348 | 60 | ||
| BC075664.1 | - | 3 - 4 | 670 | 30604610 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc3l4
Genomic Map