Assay Details
| Gene Symbol: P4htm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028944.3 | NP_083220.3 | 3 - 4 | 664 | 75 | |
| GenBank mRNA | AK016685.1 | - | 3 - 4 | 638 | 75 | |
| AK046783.1 | - | 3 - 4 | 676 | 75 | ||
| AK047714.1 | - | 3 - 4 | 664 | 75 | ||
| AK161992.1 | - | 3 - 4 | 640 | 75 | ||
| BC111871.1 | - | 3 - 4 | 664 | 40057593 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

P4htm
Genomic Map