Assay Details
| Gene Symbol: Kbtbd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278671.1 | NP_001265600.1 | 1 - 2 | 54 | 106 | |
| GenBank mRNA | AK014702.1 | - | 1 - 2 | 51 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014702.1, NM_001278671.1 transcripts.
Set Membership: |
|

Kbtbd12
Genomic Map