| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10579309 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10579310 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10579312 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10579311 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10579308 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1430258_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1428180_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10579307 |
- |
- |
| Mouse Genome 430 2.0 Array |
1428180_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1428180_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1430258_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1430258_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0800018194.mm.1 |
NM_029366.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018194.mm.1 |
NM_029366.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018194.mm.1 |
TC0800003245.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018199.mm.1 |
NM_029366.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018199.mm.1 |
NM_029366.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018199.mm.1 |
TC0800003245.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018200.mm.1 |
NM_029366.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018200.mm.1 |
NM_029366.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018200.mm.1 |
TC0800003245.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018196.mm.1 |
NM_029366.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018196.mm.1 |
NM_029366.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018196.mm.1 |
TC0800003245.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018195.mm.1 |
NM_029366.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018195.mm.1 |
NM_029366.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018195.mm.1 |
TC0800003245.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018201.mm.1 |
NM_029366.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018201.mm.1 |
NM_029366.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018201.mm.1 |
TC0800003245.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1430258_at |
- |
- |
| Mouse Expression Set 430 |
1430258_PM_at |
- |
- |
| Mouse Expression Set 430 |
1428180_PM_at |
- |
- |
| Mouse Expression Set 430 |
1428180_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1430258_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1430258_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1428180_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1428180_PM_at |
- |
- |