Assay Details
| Gene Symbol: Bcl2l12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029410.3 | NP_083686.1 | 5 - 6 | 554 | 67 | |
| GenBank mRNA | AK017362.1 | - | 5 - 6 | 555 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcl2l12
Genomic Map