Assay Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029522.2 | NP_083798.2 | 10 - 11 | 1698 | 106 | |
| GenBank mRNA | AK040996.1 | - | 10 - 11 | 1587 | 106 | |
| AK143552.1 | - | 10 - 11 | 1699 | 106 | ||
| AY081187.1 | - | 10 - 11 | 1542 | 106 | ||
| BC021308.1 | - | 9 - 10 | 1253 | 5007832 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpsm2
Genomic Map